Strychnin
Lieferant: EHRENSTORFER
C16980000
EHERC16980000EA
42
EUR
EHERC16980000
Strychnin
Strychnin
Formel:
C₂₁H₂₂N₂O₂ |
MDL:
MFCD00005941 CAS-Nummer: 57-24-9 EINECS: 200-319-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...