Sudan II
Lieferant: EHRENSTORFER
C16986102
EHERC16986102EA
94
EUR
EHERC16986102
Sudan II
Sudan II
Formel:
C₁₈H₁₆N₂O |
MDL:
MFCD00003896 CAS-Nummer: 3118-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...