Sudan IV
Lieferant: EHRENSTORFER
|
Achtung
|
C16986104
EHERC16986104EA
94
EUR
EHERC16986104
Sudan IV
Sudan IV
Formel:
C₂₄H₂₀N₄O Schmelzpunkt: 188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003893 CAS-Nummer: 85-83-6 EINECS: 201-635-8 Merck Index: 12,08538 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...