Sudanschwarz B
Lieferant: EHRENSTORFER
C16986110
EHERC16986110EA
103
EUR
EHERC16986110
Sudanschwarz B
Sudanschwarz B
Formel:
C₂₉H₂₄N₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006919 CAS-Nummer: 4197-25-5 EINECS: 224-087-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...