Sudanorange G

Lieferant: EHRENSTORFER

Synonyme: 4-(Phenylazo)resorcin, Solvent Orange 1

C16986115
EHERC16986115EA 49 EUR
EHERC16986115
Sudanorange G
Sudan Orange G
Formel: C₁₂H₁₀N₂O₂
Molecular Weight: 214,22 g/mol
Siedepunkt: 371 °C (1013 hPa)
Schmelzpunkt: 170 °C
MDL: MFCD00002275
CAS-Nummer: 2051-85-6
EINECS: 218-131-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR