Sudanrot G

Lieferant: EHRENSTORFER

Synonyme: 1-[(2-Methoxyphenyl)azo]-2-naphthol

C16986127
EHERC16986127EA 48 EUR
EHERC16986127
Sudanrot G
Sudanrot G
Formel: C₁₇H₁₄N₂O₂
Molecular Weight: 278,31 g/mol
Schmelzpunkt: 225 °C
MDL: MFCD00046377
CAS-Nummer: 1229-55-6
EINECS: 214-968-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR