Sulfachlorpyridazin
Lieferant: EHRENSTORFER
Synonyme:
4-Amino-N-(6-chloropyridazin-3-yl)benzenesulphonamide
C16990100
EHERC16990100EA
51
EUR
EHERC16990100
Sulfachlorpyridazin
Sulfachlorpyridazin
Formel:
C₁₀H₉ClN₄O₂S Molecular Weight: 284,7221 g/mol |
MDL:
MFCD00057371 CAS-Nummer: 80-32-0 EINECS: 201-269-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...