Sulfamethoxypyridazin
Lieferant: EHRENSTORFER
Synonyme:
4-Amino-N-(6-methoxypyridazin-3-yl)benzenesulphonamide
C16998150
EHERC16998150EA
51
EUR
EHERC16998150
Sulfamethoxypyridazin
Sulfamethoxypyridazin
Formel:
C₁₁H₁₂N₄O₃S Molecular Weight: 280,3 g/mol |
MDL:
MFCD00057372 CAS-Nummer: 80-35-3 EINECS: 201-272-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...