Sulfolan

Lieferant: EHRENSTORFER
Achtung

Synonyme: Thiacyclopentan-1,1-dioxid, Tetrahydrothiophen-1,1-dioxid, Tetramethylensulfon

C17008500
EHERC17008500EA 54 EUR
EHERC17008500
Sulfolan
Sulfolan
Formel: C₄H₈O₂S
Molecular Weight: 120,17 g/mol
MDL: MFCD00005484
CAS-Nummer: 126-33-0
EINECS: 204-783-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR