2,4,5-T (2,4,5-Trichlorphenoxyessigsäure)
Lieferant: EHRENSTORFER
C17100000
EHERC17100000EA
67
EUR
EHERC17100000
2,4,5-T (2,4,5-Trichlorphenoxyessigsäure)
2,4,5-T (2,4,5-Trichlorphenoxyessigsäure)
Formel:
C₈H₅Cl₃O₃ Molecular Weight: 255.48 g/mol |
MDL:
MFCD00004301 CAS-Nummer: 93-76-5 EINECS: 202-273-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...