2,4,5-T-2-Ethylhexylester (2,4,5-Trichlorphenoxyessigsäure-2-ethylhexylester)

Lieferant: EHRENSTORFER

C17110000
EHERC17110000EA 53 EUR
EHERC17110000
2,4,5-T-2-Ethylhexylester (2,4,5-Trichlorphenoxyessigsäure-2-ethylhexylester)
2,4,5-T-2-Ethylhexylester (2,4,5-Trichlorphenoxyessigsäure-2-ethylhexylester)
Formel: C₁₆H₂₁Cl₃O₃
Molecular Weight: 367.7 g/mol
CAS-Nummer: 1928-47-8
EINECS: 217-675-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR