2,4,5-T-Methylester (2,4,5-Trichlorphenoxyessigsäuremethylester)

Lieferant: EHRENSTORFER

C17125000
EHERC17125000EA 58 EUR
EHERC17125000
2,4,5-T-Methylester (2,4,5-Trichlorphenoxyessigsäuremethylester)
2,4,5-T-Methylester (2,4,5-Trichlorphenoxyessigsäuremethylester)
Formel: C₉H₇Cl₃O₃
Molecular Weight: 269.51 g/mol
CAS-Nummer: 1928-37-6
EINECS: 217-669-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR