Organic reference standard, tebuconazole, Dr. Ehrenstorfer
Lieferant: EHRENSTORFER
C17178700
EHERC17178700EA
83
EUR
EHERC17178700
Organic reference standard, tebuconazole, Dr. Ehrenstorfer
Standards
Organische Standards
This standard solution is designed for calibration or recalibration of chromatographic systems for the determination of the specified chemical compounds concerning identity and quantification and also used as a reference material for interlabortory studies to validate analytical procedures.
Standards: Produced in accordance to ISO 17034