1-Nitro-2,3,4,5-tetrachlorbenzol

Lieferant: EHRENSTORFER

Synonyme: 2,3,4,5-Tetrachloronitrobenzene

C17364500
EHERC17364500EA 42 EUR
EHERC17364500
1-Nitro-2,3,4,5-tetrachlorbenzol
1-Nitro-2,3,4,5-tetrachlorbenzol
Formel: C₆HCl₄NO₂
Molecular Weight: 260,89 g/mol
Schmelzpunkt: 66…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007064
CAS-Nummer: 879-39-0
EINECS: 212-906-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR