2,6-Dichlorbenzalchlorid
Lieferant: EHRENSTORFER
|
Gefahr
|
Synonyme:
α,α,2,6-Tetrachlortoluol
C17381000
EHERC17381000EA
40
EUR
EHERC17381000
2,6-Dichlorbenzalchlorid
2,6-Dichlorbenzalchlorid
Formel:
C₇H₄Cl₄ Molecular Weight: 229,92 g/mol Siedepunkt: 250 °C (1013 hPa) |
MDL:
MFCD00016349 CAS-Nummer: 81-19-6 EINECS: 201-332-0 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...