Tetrapentylzinn

Lieferant: EHRENSTORFER

C17415100
EHERC17415100EA 111 EUR
EHERC17415100
Tetrapentylzinn
Tetrapentylzinn
Formel: C₂₀H₄₄Sn
Molecular Weight: 403.28 g/mol
CAS-Nummer: 3765-65-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR