Theobromin

Lieferant: EHRENSTORFER
Achtung

Synonyme: 3,7-Dimethylxanthin, 3,7-Dimethyl-3,7-dihydro-2H-purin-2,6-dion, 3,7-Dimethyl-2,6(1H,3H)-purindion, 2,6-Dihydroxy-3,7-dimethylpurin, 3,7-Dimethyl-3,7-dihydro-1H-purin-2,6-dion, 3,7-Dimethyl-1H,3H-purin-2,6-dion, 3,7-Dihydro-3,7-dimethyl-1H-purin-2,6-dion

C17445900
EHERC17445900EA 0 EUR
EHERC17445900
Theobromin
Theobromin
Formel: C₇H₈N₄O₂
Molecular Weight: 180,17 g/mol
Schmelzpunkt: 357 °C
Lagertemperatur: Room Temperature
MDL: MFCD00022830
CAS-Nummer: 83-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR