Thiamin Hydrochlorid

Lieferant: EHRENSTORFER

Synonyme: Thiaminiumchlorid, Antiberiberifaktor Hydrochlorid, antineuritisches Vitamin Hydrochlorid, Aneurin Hydrochlorid, Thiaminchlorid Hydrochlorid, Vitamin B1 Hydrochlorid

C17455000
EHERC17455000EA 46 EUR
EHERC17455000
Thiamin Hydrochlorid
Thiamin Hydrochlorid
Formel: C₁₂H₁₈Cl₂N₄OS
Molecular Weight: 337.27 g/mol
Schmelzpunkt: 252
Dichte: 1.401 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012780
CAS-Nummer: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR