Thiodicarb
Lieferant: EHRENSTORFER
C17490000
EHERC17490000EA
77
EUR
EHERC17490000
Thiodicarb
Thiodicarb
Formel:
C₁₀H₁₈N₄O₄S₃ |
MDL:
MFCD00145401 CAS-Nummer: 59669-26-0 EINECS: 261-848-7 UN: 2811 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...