2-Thioxo-2,3-dihydro-4(1H)-pyrimidinon

Lieferant: EHRENSTORFER

Synonyme: 4-Hydroxy-2-mercaptopyrimidin

C17561500
EHERC17561500EA 42 EUR
EHERC17561500
2-Thioxo-2,3-dihydro-4(1H)-pyrimidinon
2-Thiouracil
Formel: C₄H₄N₂OS
Molecular Weight: 128,15 g/mol
Schmelzpunkt: 340 °C
MDL: MFCD00006039
CAS-Nummer: 141-90-2
EINECS: 205-508-8
Merck Index: 12,09504

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR