DL-Threonin
Lieferant: EHRENSTORFER
C17575000
EHERC17575000EA
98
EUR
EHERC17575000
DL-Threonin
DL-Threonin
Formel:
C₄H₉NO₃ Molecular Weight: 119,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063722 CAS-Nummer: 80-68-2 EINECS: 201-300-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...