Tolbutamid
Lieferant: EHRENSTORFER
Synonyme:
N-(Butylcarbamoyl)-4-methylbenzenesulphonamide
C17589900
EHERC17589900EA
49
EUR
EHERC17589900
Tolbutamid
Tolbutamid
Formel:
C₁₂H₁₈N₂O₃S Molecular Weight: 270.35 g/mol |
MDL:
MFCD00027169 CAS-Nummer: 64-77-7 EINECS: 200-594-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...