Triazoxid
Lieferant: EHRENSTORFER
C17660000
EHERC17660000EA
58
EUR
EHERC17660000
Triazoxid
Triazoxid
Formel:
C₁₀H₆ClN₅O |
CAS-Nummer:
72459-58-6 EINECS: 276-668-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...