Trichloronat (O-Ethyl-O-2,4,5-trichlorphenylethylthiophosphonat)

Lieferant: EHRENSTORFER

C17750000
EHERC17750000EA 110 EUR
EHERC17750000
Trichloronat (O-Ethyl-O-2,4,5-trichlorphenylethylthiophosphonat)
Trichloronat (O-Ethyl-O-2,4,5-trichlorphenylethylthiophosphonat)
Formel: C₁₀H₁₂Cl₃O₂PS
CAS-Nummer: 327-98-0
EINECS: 206-326-1
UN: 3278
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR