1,3,5-Trichlor-2-nitrobenzol
Lieferant: EHRENSTORFER
Synonyme:
1-Nitro-2,4,6-trichlorbenzol, 2,4,6-Trichlornitrobenzol
C17761300
EHERC17761300EA
38
EUR
EHERC17761300
1,3,5-Trichlor-2-nitrobenzol
1,3,5-Trichlor-2-nitrobenzol
Formel:
C₆H₂Cl₃NO₂ Molecular Weight: 226,45 g/mol Schmelzpunkt: 71 °C |
MDL:
MFCD00014690 CAS-Nummer: 18708-70-8 EINECS: 242-518-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...