2,3,4-Trichlornitrobenzol

Lieferant: EHRENSTORFER

Synonyme: 1-Nitro-2,3,4-trichlorobenzene, 1,2,3-Trichloro-4-nitrobenzene, 4-Nitro-1,2,3-trichlorobenzene

C17763400
EHERC17763400EA 44 EUR
EHERC17763400
2,3,4-Trichlornitrobenzol
2,3,4-Trichlornitrobenzol
Formel: C₆H₂Cl₃NO₂
Molecular Weight: 226,45 g/mol
Schmelzpunkt: 55,5 °C
MDL: MFCD00007063
CAS-Nummer: 17700-09-3
EINECS: 241-705-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR