2,4,5-Trichlorphenylacetat

Lieferant: EHRENSTORFER

C17775400
EHERC17775400EA 221 EUR
EHERC17775400
2,4,5-Trichlorphenylacetat
2,4,5-Trichlorphenylacetat
Formel: C₈H₅Cl₃O₂
CAS-Nummer: 5393-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR