2,4,6-Trichlorphenylacetat

Lieferant: EHRENSTORFER

C17775500
EHERC17775500EA 83 EUR
EHERC17775500
2,4,6-Trichlorphenylacetat
2,4,6-Trichlorphenylacetat
Formel: C₈H₅Cl₃O₂
CAS-Nummer: 23399-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR