Triclosan
Lieferant: EHRENSTORFER
Synonyme:
5-Chloro-2-(2,4-dichlorophenoxy)phenol, 5-Chloro-2-(2, 4-dichlorophenoxy) phenol
C17803000
EHERC17803000EA
51
EUR
EHERC17803000
Triclosan
Triclosan
Formel:
C₁₂H₇Cl₃O₂ Molecular Weight: 289,54 g/mol |
MDL:
MFCD00800992 CAS-Nummer: 3380-34-5 EINECS: 222-182-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...