Triflumuron

Lieferant: EHRENSTORFER

C17844300
EHERC17844300EA 84 EUR
EHERC17844300
Triflumuron
Triflumuron
Formel: C₁₅H₁₀ClF₃N₂O₃
MDL: MFCD00072496
CAS-Nummer: 64628-44-0
EINECS: 264-980-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR