Trioctylzinnchlorid

Lieferant: EHRENSTORFER
Achtung

C17892000
EHERC17892000EA 124 EUR
EHERC17892000
Trioctylzinnchlorid
Trioctylzinnchlorid
Formel: C₂₄H₅₁ClSn
CAS-Nummer: 2587-76-0
EINECS: 219-969-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR