Tyramin Hydrochlorid

Lieferant: EHRENSTORFER

C17895800
EHERC17895800EA 94 EUR
EHERC17895800
Tyramin Hydrochlorid
Tyramin Hydrochlorid
Formel: C₈H₁₂ClNO
Schmelzpunkt: 270…272 °C
MDL: MFCD00012901
CAS-Nummer: 60-19-5
EINECS: 200-462-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR