Vinclozolin

Lieferant: EHRENSTORFER

C17920000
EHERC17920000EA 80 EUR
EHERC17920000
Vinclozolin
Vinclozolin
Formel: C₁₂H₉Cl₂NO₃
CAS-Nummer: 50471-44-8
EINECS: 256-599-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR