2-Chlorbiphenyl (PCB Nr. 1)

Lieferant: EHRENSTORFER

Synonyme: 1-Chlor-2-phenylbenzol, PCB Nr. 1, 2-PCB, 2-Monochlorbiphenyl

C20000100
EHERC20000100EA 29.44 EUR
EHERC20000100
2-Chlorbiphenyl (PCB Nr. 1)
2-Chlorbiphenyl (PCB Nr. 1)
Formel: C₁₂H₉Cl
Molecular Weight: 188,66 g/mol
Siedepunkt: 274 °C (1013 hPa)
Schmelzpunkt: 34 °C
Dichte: 1,1499 g/cm³ (32 °C)
CAS-Nummer: 2051-60-7
EINECS: 218-125-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR