4-Chlorbiphenyl (PCB Nr. 3)

Lieferant: EHRENSTORFER

C20000300
EHERC20000300EA 29.44 EUR
EHERC20000300
4-Chlorbiphenyl (PCB Nr. 3)
4-Chlorbiphenyl (PCB Nr. 3)
Formel: C₁₂H₉Cl
Molecular Weight: 188,66 g/mol
Siedepunkt: 291 °C (1013 hPa)
Schmelzpunkt: 77…78 °C
MDL: MFCD00013640
CAS-Nummer: 2051-62-9
EINECS: 218-127-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR