2,2'-Dichlorbiphenyl (PCB Nr. 4)

Lieferant: EHRENSTORFER

C20000400
EHERC20000400EA 68 EUR
EHERC20000400
2,2'-Dichlorbiphenyl (PCB Nr. 4)
2,2'-Dichlorbiphenyl (PCB Nr. 4)
Formel: C₁₂H₈Cl₂
Molecular Weight: 223.1 g/mol
Siedepunkt: 289
Schmelzpunkt: 59…60 °C
CAS-Nummer: 13029-08-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR