2,3-Dichlorbiphenyl (PCB Nr. 5)

Lieferant: EHRENSTORFER

Synonyme: PCB Nr. 5, 2,3-PCB, 1,2-Dichlor-3-phenylbenzol

C20000500
EHERC20000500EA 45 EUR
EHERC20000500
2,3-Dichlorbiphenyl (PCB Nr. 5)
2,3-Dichlorbiphenyl (PCB Nr. 5)
Formel: C₁₂H₈Cl₂
Molecular Weight: 223,98 g/mol
MDL: MFCD00055244
CAS-Nummer: 16605-91-7
UN: 3432
ADR: 9,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR