2,4-Dichlorbiphenyl (PCB Nr. 7)
Lieferant: EHRENSTORFER
C20000700
EHERC20000700EA
32
EUR
EHERC20000700
2,4-Dichlorbiphenyl (PCB Nr. 7)
2,4-Dichlorbiphenyl (PCB Nr. 7)
Formel:
C₁₂H₈Cl₂ Molecular Weight: 223,1 g/mol |
CAS-Nummer:
33284-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...