2,4'-Dichlorbiphenyl (PCB Nr. 8)

Lieferant: EHRENSTORFER

C20000800
EHERC20000800EA 101 EUR
EHERC20000800
2,4'-Dichlorbiphenyl (PCB Nr. 8)
2,4'-Dichlorbiphenyl (PCB Nr. 8)
Formel: C₁₂H₈Cl₂
Molecular Weight: 223,1 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 34883-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR