2,6-Dichlorbiphenyl (PCB Nr. 10)

Lieferant: EHRENSTORFER

C20001000
EHERC20001000EA 45 EUR
EHERC20001000
2,6-Dichlorbiphenyl (PCB Nr. 10)
2,6-Dichlorbiphenyl (PCB Nr. 10)
Formel: C₁₂H₈Cl₂
Molecular Weight: 223,1 g/mol
CAS-Nummer: 33146-45-1
UN: 3432
ADR: 9,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR