3,4'-Dichlorbiphenyl (PCB Nr. 13)

Lieferant: EHRENSTORFER

Synonyme: 3,4'-PCB, 1-Chlor-3-(4-chlorphenyl)benzol, PCB Nr. 13

C20001300
EHERC20001300EA 96 EUR
EHERC20001300
3,4'-Dichlorbiphenyl (PCB Nr. 13)
3,4'-Dichlorbiphenyl (PCB Nr. 13)
Formel: C₁₂H₈Cl₂
Molecular Weight: 223,1 g/mol
MDL: MFCD00152402
CAS-Nummer: 2974-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR