3,5-Dichlorbiphenyl (PCB Nr. 14)
Lieferant: EHRENSTORFER
Synonyme:
PCB Nr. 14, 1,3-Dichlor-5-phenylbenzol, 3,5-PCB
C20001400
EHERC20001400EA
62
EUR
EHERC20001400
3,5-Dichlorbiphenyl (PCB Nr. 14)
3,5-Dichlorbiphenyl (PCB Nr. 14)
Formel:
C₁₂H₈Cl₂ Molecular Weight: 223,1 g/mol |
MDL:
MFCD00055256 CAS-Nummer: 34883-41-5 UN: 3432 ADR: 9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...