4,4'-Dichlorbiphenyl (PCB Nr. 15)

Lieferant: EHRENSTORFER

Synonyme: 4,4'-PCB, PCB Nr. 15, 1-Chlor-4-(4-chlorphenyl)benzol

C20001500
EHERC20001500EA 29.81 EUR
EHERC20001500
4,4'-Dichlorbiphenyl (PCB Nr. 15)
4,4'-Dichlorbiphenyl (PCB Nr. 15)
Formel: C₁₂H₈Cl₂
Molecular Weight: 223,1 g/mol
CAS-Nummer: 2050-68-2
EINECS: 218-095-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR