2,2',5-Trichlorbiphenyl (PCB Nr. 18)
Lieferant: EHRENSTORFER
C20001800
EHERC20001800EA
64
EUR
EHERC20001800
2,2',5-Trichlorbiphenyl (PCB Nr. 18)
2,2',5-Trichlorbiphenyl (PCB Nr. 18)
Formel:
C₁₂H₇Cl₃ Molecular Weight: 257.55 g/mol Schmelzpunkt: 44 Dichte: 1.1485 g/cm³ (20 °C) |
CAS-Nummer:
37680-65-2 UN: 3432 ADR: 9 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...