2,3,3'-Trichlorbiphenyl (PCB Nr. 20)

Lieferant: EHRENSTORFER

C20002000
EHERC20002000EA 60 EUR
EHERC20002000
2,3,3'-Trichlorbiphenyl (PCB Nr. 20)
2,3,3'-Trichlorbiphenyl (PCB Nr. 20)
Formel: C₁₂H₇Cl₃
Molecular Weight: 257.55 g/mol
CAS-Nummer: 38444-84-7
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR