2,3,4-Trichlorbiphenyl (PCB Nr. 21)

Lieferant: EHRENSTORFER

C20002100
EHERC20002100EA 50 EUR
EHERC20002100
2,3,4-Trichlorbiphenyl (PCB Nr. 21)
2,3,4-Trichlorbiphenyl (PCB Nr. 21)
Formel: C₁₂H₇Cl₃
Molecular Weight: 257.55 g/mol
CAS-Nummer: 55702-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR