2,4,5-Trichlorbiphenyl (PCB Nr. 29)

Lieferant: EHRENSTORFER

C20002900
EHERC20002900EA 40 EUR
EHERC20002900
2,4,5-Trichlorbiphenyl (PCB Nr. 29)
2,4,5-Trichlorbiphenyl (PCB Nr. 29)
Formel: C₁₂H₇Cl₃
Molecular Weight: 257.55 g/mol
CAS-Nummer: 15862-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR