2,2',3,3'-Tetrachlorbiphenyl (PCB Nr. 40)

Lieferant: EHRENSTORFER

C20004000
EHERC20004000EA 56 EUR
EHERC20004000
2,2',3,3'-Tetrachlorbiphenyl (PCB Nr. 40)
2,2',3,3'-Tetrachlorbiphenyl (PCB Nr. 40)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 38444-93-8
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR