2,2',3,5'-Tetrachlorbiphenyl (PCB Nr. 44)

Lieferant: EHRENSTORFER

C20004400
EHERC20004400EA 87 EUR
EHERC20004400
2,2',3,5'-Tetrachlorbiphenyl (PCB Nr. 44)
2,2',3,5'-Tetrachlorbiphenyl (PCB Nr. 44)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 41464-39-5
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR