2,2',5,5'-Tetrachlorbiphenyl (PCB Nr. 52)

Lieferant: EHRENSTORFER

C20005200
EHERC20005200EA 39 EUR
EHERC20005200
2,2',5,5'-Tetrachlorbiphenyl (PCB Nr. 52)
2,2',5,5'-Tetrachlorbiphenyl (PCB Nr. 52)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 35693-99-3
UN: 2315
ADR: 9 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR